Assay Details
Target Gene Details
Entrez Gene ID: | 55294 |
Gene Name: | F-box and WD repeat domain containing 7 |
Gene Aliases: |
AGO, CDC4, FBW6, FBW7, FBX30, FBXO30, FBXW6, SEL-10, SEL10, hAgo, hCdc4 |
Location: |
Chr.4:152321258-152536095 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FBXW7 | NM_001013415.1 | 11 | 2694 | NP_001013433.1 |
| NM_018315.4 | 11 | 2859 | NP_060785.2 | |
| NM_033632.3 | 12 | 3033 | NP_361014.1 | |
| XM_011532083.1 | 14 | 4046 | XP_011530385.1 | |
| XM_011532084.1 | 13 | 3996 | XP_011530386.1 | |
| XM_011532085.1 | 12 | 3903 | XP_011530387.1 | |
| XM_011532086.1 | 15 | 4224 | XP_011530388.1 | |
| XM_011532087.1 | 14 | 4174 | XP_011530389.1 | |
| XM_011532088.1 | 12 | 3432 | XP_011530390.1 | |
| XM_017008362.1 | 14 | 4443 | XP_016863851.1 | |
| AY008274.1 | 11 | 2696 | AAG16640.1 | |
| BC037320.2 | 11 | 2614 | AAH37320.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv516218 | Chr.4:152308716 - 152388386 on Build GRCh38 | Loss |
|
| dgv5440n100 | Chr.4:152321467 - 152392321 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map