Assay Details
Target Gene Details
Entrez Gene ID: | 474 |
Gene Name: | atonal bHLH transcription factor 1 |
Gene Aliases: |
ATH1, HATH1, MATH-1, bHLHa14 |
Location: |
Chr.4:93828927-93829991 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATOH1 | NM_005172.1 | 1 | 903 | NP_005163.1 |
| BC069145.1 | 1 | 904 | AAH69145.1 | |
| BC069578.1 | 1 | 898 | AAH69578.1 | |
| BC069594.1 | 1 | 903 | AAH69594.1 | |
| BC069604.1 | 1 | 903 | AAH69604.1 | |
| BC113623.1 | 1 | 939 | ||
| BC113625.1 | 1 | 939 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv997789 | Chr.4:92938642 - 94028074 on Build GRCh38 | Gain |
|
| nsv821636 | Chr.4:93518485 - 94195439 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map