Assay Details
Target Gene Details
Entrez Gene ID: | 4946 |
Gene Name: | ornithine decarboxylase antizyme 1 |
Gene Aliases: |
AZI, OAZ |
Location: |
Chr.19:2269486-2273488 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OAZ1 | NM_001301020.1 | NP_001287949.1 | ||
| NM_004152.3 | NP_004143.1 | |||
| AK307221.1 | ||||
| AL542477.3 | ||||
| BI667301.1 | ||||
| BM423450.1 | ||||
| D78361.1 | BAA11373.1 | |||
| D87914.1 | BAA13497.1 | |||
| U09202.1 | AAA82155.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv6218n54 | Chr.19:2221793 - 2336009 on Build GRCh38 | Loss |
|
| nsv578383 | Chr.19:2214058 - 2285197 on Build GRCh38 | Loss |
|
| nsv833707 | Chr.19:2187092 - 2372223 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map