Assay Details
Target Gene Details
Entrez Gene ID: | 1627 |
Gene Name: | drebrin 1 |
Gene Aliases: |
D0S117E |
Location: |
Chr.5:177456612-177480368 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DBN1 | NM_004395.3 | 14 | 2379 | NP_004386.2 |
| NM_080881.2 | 15 | 2495 | NP_543157.1 | |
| XM_005265827.3 | 15 | 2464 | XP_005265884.1 | |
| XM_006714826.2 | 15 | 2354 | XP_006714889.1 | |
| XM_011534446.2 | 16 | 2636 | XP_011532748.1 | |
| XM_011534447.2 | 15 | 3133 | XP_011532749.1 | |
| XM_017009139.1 | 15 | 2635 | XP_016864628.1 | |
| XM_017009140.1 | 14 | 2488 | XP_016864629.1 | |
| AK094125.1 | 13 | 2425 | ||
| AK225523.1 | 14 | 2320 | ||
| AW950551.1 | ||||
| BC000283.1 | 14 | 2303 | AAH00283.1 | |
| BC007281.1 | 14 | 2303 | AAH07281.1 | |
| BC007567.1 | AAH07567.1 | |||
| D17530.1 | 14 | 2304 | BAA04480.1 | |
| U00802.1 | 14 | 2279 | AAA16256.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv471057 | Chr.5:177310308 - 177469411 on Build GRCh38 | Loss |
|
| nsv1161313 | Chr.5:177356552 - 177463415 on Build GRCh38 | Duplication |
|
| dgv733n27 | Chr.5:177357511 - 177469411 on Build GRCh38 | Loss |
|
| nsv515805 | Chr.5:177432847 - 177469411 on Build GRCh38 | Loss |
|
| nsv1161311 | Chr.5:177305104 - 177546474 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map