Assay Details
Target Gene Details
Entrez Gene ID: | 7433 |
Gene Name: | vasoactive intestinal peptide receptor 1 |
Gene Aliases: |
HVR1, II, PACAP-R-2, PACAP-R2, RDC1, V1RG, VAPC1, VIP-R-1, VIPR, VIRG, VPAC1, VPAC1R, VPCAP1R |
Location: |
Chr.3:42489299-42537573 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VIPR1 | NM_001251882.1 | NP_001238811.1 | ||
| NM_001251883.1 | NP_001238812.1 | |||
| NM_001251884.1 | NP_001238813.1 | |||
| NM_001251885.1 | NP_001238814.1 | |||
| NM_004624.3 | NP_004615.2 | |||
| XM_005265437.2 | XP_005265494.1 | |||
| XM_005265438.3 | XP_005265495.1 | |||
| XM_005265439.2 | XP_005265496.1 | |||
| XM_011534079.1 | XP_011532381.1 | |||
| XM_011534080.2 | XP_011532382.1 | |||
| AK056819.1 | ||||
| AK293548.1 | ||||
| AK294609.1 | ||||
| AK298115.1 | ||||
| AK314334.1 | ||||
| BC064424.1 | AAH64424.1 | |||
| EF577396.1 | ||||
| L13288.1 | AAA36805.1 | |||
| L20295.1 | AAA36802.1 | |||
| X75299.1 | CAA53046.1 |
Target Gene Details
Entrez Gene ID: | 100874007 |
Gene Name: | VIPR1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:42530896-42532606 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VIPR1-AS1 | NR_046654.1 | |||
| AA620928.1 |
More Information
Set Membership: |
|

Genomic Map