Assay Details
Target Gene Details
Entrez Gene ID: | 55255 |
Gene Name: | WD repeat domain 41 |
Gene Aliases: |
MSTP048 |
Location: |
Chr.5:77426652-77620611 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WDR41 | NM_018268.3 | NP_060738.2 | ||
| XM_005248551.4 | XP_005248608.1 | |||
| XM_005248552.3 | XP_005248609.1 | |||
| XM_011543505.2 | XP_011541807.1 | |||
| XM_011543507.2 | XP_011541809.1 | |||
| XM_017009606.1 | XP_016865095.1 | |||
| AF115511.1 | AAO06948.1 | |||
| AK001766.1 | BAA91895.1 | |||
| AK021855.1 | BAB13915.1 | |||
| AK023977.1 | BAB14749.1 | |||
| AK293316.1 | ||||
| AK295249.1 | ||||
| AK300058.1 | ||||
| AK304329.1 | ||||
| AL834138.1 | CAD38853.2 | |||
| BC040241.1 | AAH40241.1 | |||
| KU178690.1 | ||||
| KU178691.1 |

Genomic Map