Assay Details
Target Gene Details
Entrez Gene ID: | 221044 |
Gene Name: | upper zone of growth plate and cartilage matrix associated |
Gene Aliases: |
C10orf49, GRP, GRP/UCMA |
Location: |
Chr.10:13221766-13234328 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UCMA | NM_001303118.1 | 4 | 555 | NP_001290047.1 |
| NM_001303119.1 | 3 | 489 | NP_001290048.1 | |
| NM_145314.2 | 5 | 651 | NP_660357.2 | |
| AA628504.1 | 1 | 178 | ||
| BC018068.1 | 5 | 654 | AAH18068.2 |
More Information
Set Membership: |
|

Genomic Map