Assay Details
Target Gene Details
Entrez Gene ID: | 4720 |
Gene Name: | NADH:ubiquinone oxidoreductase core subunit S2 |
Gene Aliases: |
CI-49 |
Location: |
Chr.1:161197377-161214395 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDUFS2 | NM_001166159.1 | NP_001159631.1 | ||
| NM_004550.4 | NP_004541.1 | |||
| XM_005245208.2 | XP_005245265.1 | |||
| XM_005245209.1 | XP_005245266.1 | |||
| XM_017001357.1 | XP_016856846.1 | |||
| AF013160.1 | AAC34362.1 | |||
| AF050640.1 | AAC27453.1 | |||
| AK222618.1 | BAD96338.1 | |||
| AK301641.1 | ||||
| AK309871.1 | ||||
| AK314807.1 | ||||
| AK315977.1 | ||||
| BC000170.2 | AAH00170.1 | |||
| BC001456.1 | AAH01456.1 | |||
| BC008868.2 | AAH08868.1 | |||
| Z36812.1 |
More Information
Set Membership: |
|

Genomic Map