Assay Details
Target Gene Details
Entrez Gene ID: | 27351 |
Gene Name: | desumoylating isopeptidase 1 |
Gene Aliases: |
D15Wsu75e, DESI2, DJ347H13.4, DeSI-1, FAM152B, PPPDE2 |
Location: |
Chr.22:41598028-41621077 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DESI1 | NM_015704.2 | 6 | 873 | NP_056519.1 |
| AF038183.1 | 6 | 831 | ||
| AK090725.1 | ||||
| BX403547.2 | ||||
| BX478162.1 | 1 | 219 | ||
| CR456458.1 | CAG30344.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1055166 | Chr.22:41536399 - 41622240 on Build GRCh38 | Gain |
|
| nsv834209 | Chr.22:41563516 - 41746056 on Build GRCh38 | Loss |
|
| nsv524585 | Chr.22:40993505 - 41794305 on Build GRCh38 | Loss |
|
| nsv3639 | Chr.22:41571809 - 41602871 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map