Assay Details
Target Gene Details
Entrez Gene ID: | 5570 |
Gene Name: | protein kinase (cAMP-dependent, catalytic) inhibitor beta |
Gene Aliases: |
PRKACN2 |
Location: |
Chr.6:122471917-122726373 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PKIB | NM_001270393.1 | NP_001257322.1 | ||
| NM_001270394.1 | NP_001257323.1 | |||
| NM_181794.2 | NP_861459.1 | |||
| AJ420562.1 | ||||
| BG507119.1 | ||||
| BM802552.1 | ||||
| BM969901.1 | ||||
| CR749456.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759467 | Chr.6:122402790 - 122562992 on Build GRCh38 | Loss |
|
| nsv1018215 | Chr.6:122467666 - 122706716 on Build GRCh38 | Loss |
|
| nsv1161448 | Chr.6:122422593 - 122476539 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map