Assay Details
Target Gene Details
Entrez Gene ID: | 8403 |
Gene Name: | SRY-box 14 |
Gene Aliases: |
SOX28 |
Location: |
Chr.3:137764292-137766334 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SOX14 | NM_004189.3 | 1 | 1269 | NP_004180.1 |
| BC106729.1 | AAI06730.1 | |||
| BC106730.1 | AAI06731.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv955354 | Chr.3:137759259 - 137771558 on Build GRCh38 | Deletion |
|
| nsv1142374 | Chr.3:137757320 - 137778868 on Build GRCh38 | Tandem Duplication |
|
| nsv428422 | Chr.3:137344347 - 138686784 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map