Assay Details
Target Gene Details
Entrez Gene ID: | 84687 |
Gene Name: | protein phosphatase 1 regulatory subunit 9B |
Gene Aliases: |
PPP1R6, PPP1R9, SPINO, Spn |
Location: |
Chr.17:50133735-50151027 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP1R9B | NM_032595.4 | 10 | 3226 | NP_115984.3 |
| AK054907.1 | 8 | 1699 | ||
| AL157449.1 | 1 | 199 | ||
| AL713642.1 | 10 | 2033 | CAD28455.2 | |
| BC016162.1 | 8 | 1713 | AAH16162.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952339 | Chr.17:50115237 - 50222139 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map