Assay Details
Target Gene Details
Entrez Gene ID: | 3576 |
Gene Name: | C-X-C motif chemokine ligand 8 |
Gene Aliases: |
GCP-1, GCP1, IL8, LECT, LUCT, LYNAP, MDNCF, MONAP, NAF, NAP-1, NAP1 |
Location: |
Chr.4:73740506-73743716 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CXCL8 | NM_000584.3 | 4 | 490 | NP_000575.1 |
| AK131067.1 | 4 | 427 | ||
| AK311874.1 | ||||
| BC013615.1 | 4 | 377 | AAH13615.1 | |
| BG497712.1 | 4 | 491 | ||
| M17017.1 | 4 | 427 | AAA35611.1 | |
| M26383.1 | 4 | 411 | AAA36323.1 | |
| Y00787.1 | 4 | 438 | CAA68742.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1001725 | Chr.4:73488333 - 73995207 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map