Assay Details
Target Gene Details
Entrez Gene ID: | 27107 |
Gene Name: | zinc finger and BTB domain containing 11 |
Gene Aliases: |
ZNF-U69274, ZNF913 |
Location: |
Chr.3:101649439-101677144 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZBTB11 | NM_014415.3 | 11 | 3387 | NP_055230.2 |
| XM_011512689.2 | 11 | 3095 | XP_011510991.1 | |
| XM_011512690.2 | 9 | 4772 | XP_011510992.1 | |
| AB208862.1 | 8 | 2100 | BAD92099.1 | |
| AK023488.1 | 3 | 839 | ||
| AK312859.1 | ||||
| BC111700.1 | 11 | 3294 | AAI11701.1 | |
| U69274.1 | 11 | 3318 | AAD00172.1 |
More Information
Set Membership: |
|

Genomic Map