Assay Details
Target Gene Details
Entrez Gene ID: | 80821 |
Gene Name: | DDHD domain containing 1 |
Gene Aliases: |
PA-PLA1, PAPLA1, SPG28 |
Location: |
Chr.14:53036740-53153328 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDHD1 | NM_001160147.1 | 13 | 3141 | NP_001153619.1 |
| NM_001160148.1 | 13 | 3204 | NP_001153620.1 | |
| NM_030637.2 | 12 | 3120 | NP_085140.2 | |
| XM_005268102.2 | 14 | 3295 | XP_005268159.1 | |
| XM_005268103.2 | 14 | 3214 | XP_005268160.1 | |
| XM_005268105.2 | 12 | 3064 | XP_005268162.1 | |
| XM_011537188.2 | 15 | 3316 | XP_011535490.1 | |
| XM_011537189.2 | 14 | 3232 | XP_011535491.1 | |
| XM_017021668.1 | 13 | 3211 | XP_016877157.1 | |
| XM_017021669.1 | 11 | 2980 | XP_016877158.1 | |
| AB051492.1 | 9 | 3481 | BAB21796.1 | |
| AK058137.1 | 10 | 1823 | BAB71679.1 | |
| BC018014.1 | 4 | 852 | AAH18014.1 | |
| BC030703.2 | 12 | 3074 | AAH30703.1 | |
| DQ315474.1 | 13 | 2633 | ABC43196.1 | |
| GU376468.1 | 14 | 2719 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832797 | Chr.14:52932853 - 53113976 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map