Assay Details
Target Gene Details
Entrez Gene ID: | 9290 |
Gene Name: | G protein-coupled receptor 55 |
Gene Aliases: |
LPIR1 |
Location: |
Chr.2:230907318-230961269 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR55 | NM_005683.3 | 2 | 486 | NP_005674.2 |
| XM_005246952.3 | 2 | 746 | XP_005247009.1 | |
| XM_011512175.2 | 2 | 922 | XP_011510477.1 | |
| XM_011512176.2 | 2 | 505 | XP_011510478.1 | |
| AK292223.1 | 2 | 631 | ||
| BC032694.1 | 2 | 486 | AAH32694.1 | |
| CR541776.1 | 1 | 293 | CAG46575.1 | |
| FJ348259.1 | 1 | 293 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470522 | Chr.2:230867985 - 230943576 on Build GRCh38 | Loss |
|
| nsv834565 | Chr.2:230697957 - 230920661 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map