Assay Details
Target Gene Details
Entrez Gene ID: | 83943 |
Gene Name: | inner mitochondrial membrane peptidase subunit 2 |
Gene Aliases: |
IMMP2L-IT1, IMP2, IMP2-LIKE |
Location: |
Chr.7:110662645-111562531 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IMMP2L | NM_001244606.1 | NP_001231535.1 | ||
| NM_032549.3 | NP_115938.1 | |||
| XM_005250630.3 | XP_005250687.1 | |||
| XM_011516605.2 | XP_011514907.1 | |||
| XM_011516608.2 | XP_011514910.1 | |||
| XM_011516609.2 | XP_011514911.1 | |||
| XM_011516613.2 | XP_011514915.1 | |||
| XM_017012699.1 | XP_016868188.1 | |||
| XM_017012700.1 | XP_016868189.1 | |||
| XM_017012701.1 | XP_016868190.1 | |||
| XM_017012702.1 | XP_016868191.1 | |||
| XM_017012703.1 | XP_016868192.1 | |||
| XM_017012704.1 | XP_016868193.1 | |||
| XM_017012705.1 | XP_016868194.1 | |||
| AF359563.1 | AAK52905.1 | |||
| AK311968.1 | ||||
| BC008497.1 | AAH08497.1 |
Target Gene Details
Entrez Gene ID: | 54674 |
Gene Name: | leucine rich repeat neuronal 3 |
Gene Aliases: |
FIGLER5, NLRR-3, NLRR3 |
Location: |
Chr.7:111091006-111125454 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRRN3 | NM_001099658.1 | 3 | 1515 | NP_001093128.1 |
| NM_001099660.1 | 4 | 1622 | NP_001093130.1 | |
| NM_018334.4 | 2 | 1433 | NP_060804.3 | |
| AB060967.1 | 1 | 882 | BAB47184.1 | |
| AF134481.1 | 1 | 913 | AAP97258.1 | |
| AK001991.1 | 4 | 1534 | BAA92025.1 | |
| AK292556.1 | 2 | 1022 | ||
| AL442092.1 | 1 | 838 | CAC09450.1 | |
| AY358280.1 | 3 | 1055 | AAQ88647.1 | |
| BC035133.2 | 2 | 1312 | AAH35133.1 | |
| CR457397.1 | 1 | 576 | CAG33678.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1024477 | Chr.7:111019950 - 111304421 on Build GRCh38 | Loss |
|
| nsv608154 | Chr.7:111048937 - 111305445 on Build GRCh38 | Loss |
|
| esv2759554 | Chr.7:110883242 - 111147793 on Build GRCh38 | Loss |
|
| nsv608080 | Chr.7:109461248 - 111666297 on Build GRCh38 | Gain |
|
| nsv608155 | Chr.7:111082426 - 111711456 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map