Assay Details
Target Gene Details
Entrez Gene ID: | 57217 |
Gene Name: | tetratricopeptide repeat domain 7A |
Gene Aliases: |
GIDID, MINAT, TTC7 |
Location: |
Chr.2:46915931-47076136 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC7A | NM_001288951.1 | NP_001275880.1 | ||
| NM_001288953.1 | NP_001275882.1 | |||
| NM_001288955.1 | NP_001275884.1 | |||
| NM_020458.3 | NP_065191.2 | |||
| XM_005264439.3 | XP_005264496.1 | |||
| XM_011532998.2 | XP_011531300.1 | |||
| XM_011532999.2 | XP_011531301.1 | |||
| XM_017004524.1 | XP_016860013.1 | |||
| XM_017004525.1 | XP_016860014.1 | |||
| XM_017004526.1 | XP_016860015.1 | |||
| XM_017004527.1 | XP_016860016.1 | |||
| XM_017004528.1 | XP_016860017.1 | |||
| XM_017004529.1 | XP_016860018.1 | |||
| XM_017004530.1 | XP_016860019.1 | |||
| AB032966.2 | ||||
| AK056464.1 | ||||
| AL834383.1 | CAD39046.2 | |||
| BC111487.1 | AAI11488.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524970 | Chr.2:46935600 - 47074956 on Build GRCh38 | Loss |
|
| nsv457352 | Chr.2:46777649 - 47098834 on Build GRCh38 | Loss |
|
| nsv1004828 | Chr.2:46829710 - 47587343 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map