Assay Details
Target Gene Details
Entrez Gene ID: | 4720 |
Gene Name: | NADH:ubiquinone oxidoreductase core subunit S2 |
Gene Aliases: |
CI-49 |
Location: |
Chr.1:161197377-161214395 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDUFS2 | NM_001166159.1 | 1 | 91 | NP_001159631.1 |
| NM_004550.4 | 2 | 300 | NP_004541.1 | |
| XM_005245208.2 | 2 | 202 | XP_005245265.1 | |
| XM_017001357.1 | 1 | 566 | XP_016856846.1 | |
| BC008868.2 | 2 | 300 | AAH08868.1 |
More Information
Set Membership: |
|

Genomic Map