Assay Details
Target Gene Details
Entrez Gene ID: | 9517 |
Gene Name: | serine palmitoyltransferase long chain base subunit 2 |
Gene Aliases: |
HSN1C, LCB2, LCB2A, NSAN1C, SPT2, hLCB2a |
Location: |
Chr.14:77505997-77616767 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPTLC2 | NM_004863.3 | 12 | 4761 | NP_004854.1 |
| AB011098.1 | 12 | 4761 | ||
| BG024616.1 | 1 | 508 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832835 | Chr.14:77440363 - 77628864 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map