Assay Details
Target Gene Details
Entrez Gene ID: | 115 |
Gene Name: | adenylate cyclase 9 |
Gene Aliases: |
AC9, ACIX |
Location: |
Chr.16:3953387-4116185 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADCY9 | NM_001116.3 | 11 | 4423 | NP_001107.2 |
| XM_005255079.3 | 11 | 4305 | XP_005255136.1 | |
| XM_011522353.2 | XP_011520655.1 | |||
| AB011092.3 | 11 | 4264 | ||
| AF036927.1 | 10 | 3900 | AAC24201.1 | |
| AJ133123.1 | 11 | 4422 | CAB65084.1 | |
| AU141090.1 | 1 | 514 | ||
| BC136657.1 | 10 | 3940 | ||
| BC136658.1 | 10 | 3940 | ||
| BC151207.1 | 11 | 4259 | ||
| BC151229.1 | 11 | 4264 | ||
| DQ005545.1 | 10 | 3884 | AAY21237.1 | |
| DQ008441.1 | 10 | 3884 | AAY27880.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv21490 | Chr.16:3825338 - 4270692 on Build GRCh38 | Gain+Loss |
|
| nsv833131 | Chr.16:3955868 - 4135556 on Build GRCh38 | Loss |
|
| nsv1038196 | Chr.16:3917410 - 4153984 on Build GRCh38 | Gain |
|
| nsv1039014 | Chr.16:3965728 - 4070295 on Build GRCh38 | Gain |
|
| nsv833130 | Chr.16:3810372 - 3975509 on Build GRCh38 | Gain |
|
| esv3570176 | Chr.16:3960729 - 4005988 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map