Assay Details
Target Gene Details
Entrez Gene ID: | 10809 |
Gene Name: | StAR related lipid transfer domain containing 10 |
Gene Aliases: |
CGI-52, NY-CO-28, PCTP2, SDCCAG28 |
Location: |
Chr.11:72754729-72793705 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STARD10 | NM_006645.2 | NP_006636.2 | ||
| AF151810.1 | AAD34047.1 | |||
| AK095668.1 | ||||
| AK315255.1 | ||||
| BC007919.2 | AAH07919.1 | |||
| BC014033.2 | AAH14033.1 |
More Information
Set Membership: |
|

Genomic Map