Assay Details
Target Gene Details
Entrez Gene ID: | 101929464 |
Gene Name: | uncharacterized LOC101929464 |
Gene Aliases: |
- |
Location: |
Chr.1:33350352-33363245 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101929464 | NR_125978.1 | |||
| AW072537.1 |
Target Gene Details
Entrez Gene ID: | 1912 |
Gene Name: | polyhomeotic homolog 2 |
Gene Aliases: |
EDR2, HPH2, PH2 |
Location: |
Chr.1:33323623-33431017 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PHC2 | NM_198040.2 | 7 | 1404 | NP_932157.1 |
| XM_011540876.2 | 8 | 1939 | XP_011539178.1 | |
| XM_011540877.2 | 8 | 1939 | XP_011539179.1 | |
| XM_011540878.2 | 8 | 1939 | XP_011539180.1 | |
| XM_017000513.1 | 8 | 1900 | XP_016856002.1 | |
| XM_017000514.1 | 8 | 1512 | XP_016856003.1 | |
| XM_017000515.1 | 8 | 1605 | XP_016856004.1 | |
| XM_017000516.1 | 8 | 1834 | XP_016856005.1 | |
| XM_017000517.1 | 8 | 1446 | XP_016856006.1 | |
| XM_017000518.1 | 8 | 1539 | XP_016856007.1 | |
| XM_017000519.1 | 8 | 1909 | XP_016856008.1 | |
| XM_017000520.1 | 7 | 1359 | XP_016856009.1 | |
| XM_017000521.1 | 7 | 1778 | XP_016856010.1 | |
| XM_017000522.1 | 8 | 1939 | XP_016856011.1 | |
| AJ419231.1 | 7 | 1404 | CAD11673.1 | |
| AK056429.1 | 1 | 331 | ||
| AK292905.1 | 8 | 1839 | ||
| AK294426.1 | 2 | 679 | ||
| BC130630.1 | 6 | 1317 | ||
| BC144121.1 | 6 | 1317 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834613 | Chr.1:33334605 - 33496188 on Build GRCh38 | Gain |
|
| nsv522364 | Chr.1:33266591 - 33367332 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map