Assay Details
Target Gene Details
Entrez Gene ID: | 55101 |
Gene Name: | ATP5S like |
Gene Aliases: |
- |
Location: |
Chr.19:41431318-41447822 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Entrez Gene ID: | 55101 |
Gene Name: | ATP5S like |
Gene Aliases: |
- |
Location: |
Chr.19:41431318-41447822 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |