Assay Details
Target Gene Details
Entrez Gene ID: | 6327 |
Gene Name: | sodium voltage-gated channel beta subunit 2 |
Gene Aliases: |
ATFB14 |
Location: |
Chr.11:118162804-118176622 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SCN2B | NM_004588.4 | 4 | 2298 | NP_004579.1 |
| AK094643.1 | 1 | 221 | ||
| AK308834.1 | 4 | 868 |
More Information
Set Membership: |
|

Genomic Map