Assay Details
Target Gene Details
Entrez Gene ID: | 29843 |
Gene Name: | SUMO1/sentrin specific peptidase 1 |
Gene Aliases: |
SuPr-2 |
Location: |
Chr.12:48042893-48106308 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 20 - Exon 20 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759898 | Chr.12:48035919 - 48431716 on Build GRCh38 | Gain+Loss |
|
| nsv832401 | Chr.12:47953670 - 48084216 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map