Assay Details
Target Gene Details
Entrez Gene ID: | 9039 |
Gene Name: | ubiquitin like modifier activating enzyme 3 |
Gene Aliases: |
NAE2, UBE1C, hUBA3 |
Location: |
Chr.3:69054730-69080373 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBA3 | NM_003968.3 | NP_003959.3 | ||
| NM_198195.1 | NP_937838.1 | |||
| XM_011534210.1 | XP_011532512.1 | |||
| XM_011534211.1 | XP_011532513.1 | |||
| AB012190.1 | BAA33144.1 | |||
| AF046024.1 | AAC27648.1 | |||
| AK002159.1 | ||||
| AK289392.1 | ||||
| AK298878.1 | ||||
| AK314751.1 | ||||
| AK316540.1 | ||||
| AL117566.1 | CAB55996.2 | |||
| BC022853.1 | AAH22853.1 | |||
| BG940987.1 | ||||
| CD700642.1 | ||||
| CR533537.1 | CAG38568.1 |
More Information
Set Membership: |
|

Genomic Map