Assay Details
Target Gene Details
Entrez Gene ID: | 147372 |
Gene Name: | collagen and calcium binding EGF domains 1 |
Gene Aliases: |
HKLLS1 |
Location: |
Chr.18:59430939-59698181 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCBE1 | NM_133459.3 | 11 | 2286 | NP_597716.1 |
| XM_017025556.1 | 11 | 3166 | XP_016881045.1 | |
| XM_017025557.1 | 12 | 3059 | XP_016881046.1 | |
| AB075863.1 | 11 | 2254 | ||
| AF087966.1 | ||||
| AK055243.1 | 1 | 1189 | ||
| AY358347.1 | 11 | 2232 | AAQ88713.1 | |
| BX640826.1 | 6 | 1542 | CAE45902.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs78905013] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map