Assay Details
Target Gene Details
Entrez Gene ID: | 1499 |
Gene Name: | catenin beta 1 |
Gene Aliases: |
CTNNB, MRD19, armadillo |
Location: |
Chr.3:41199451-41240448 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CTNNB1 | NM_001098209.1 | 5 | 824 | NP_001091679.1 |
| NM_001098210.1 | 5 | 824 | NP_001091680.1 | |
| NM_001904.3 | 5 | 824 | NP_001895.1 | |
| XM_005264886.2 | 5 | 838 | XP_005264943.1 | |
| XM_006712983.1 | 5 | 736 | XP_006713046.1 | |
| XM_006712984.1 | 5 | 736 | XP_006713047.1 | |
| XM_006712985.1 | 5 | 798 | XP_006713048.1 | |
| XM_017005738.1 | 5 | 838 | XP_016861227.1 | |
| AB451264.1 | 3 | 556 | ||
| AB451392.1 | 3 | 556 | ||
| AK289932.1 | 6 | 712 | ||
| AK294778.1 | 6 | 937 | ||
| AK296790.1 | 5 | 644 | ||
| AK299954.1 | 5 | 506 | ||
| AK314776.1 | 5 | 770 | ||
| BC058926.1 | 5 | 766 | AAH58926.1 | |
| X87838.1 | 5 | 770 | CAA61107.1 | |
| Z19054.1 | 5 | 756 | CAA79497.1 |
More Information
Set Membership: |
|

Genomic Map