Assay Details
Target Gene Details
Entrez Gene ID: | 55640 |
Gene Name: | feline leukemia virus subgroup C cellular receptor family member 2 |
Gene Aliases: |
C14orf58, CCT, EPV, FLVCRL14q, MFSD7C, PVHH |
Location: |
Chr.14:75578597-75648169 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FLVCR2 | NM_017791.2 | 1 | 349 | NP_060261.2 |
| AF456126.1 | AAO15528.1 | |||
| AK000378.1 | 1 | 297 | BAA91126.1 | |
| AY260572.1 | 1 | 329 | AAP86633.1 | |
| AY260577.1 | 1 | 329 | AAP86638.1 | |
| BC019087.2 | 1 | 312 | AAH19087.1 | |
| DA166212.1 | 1 | 349 |
Target Gene Details
Entrez Gene ID: | 102724153 |
Gene Name: | uncharacterized LOC102724153 |
Gene Aliases: |
- |
Location: |
Chr.14:75574901-75579588 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC102724153 | NR_110552.1 | 1 | 644 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv456342 | Chr.14:75553412 - 75579019 on Build GRCh38 | Loss |
|
| nsv1037822 | Chr.14:75468606 - 75633843 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map