Assay Details
Target Gene Details
Entrez Gene ID: | 90525 |
Gene Name: | Src homology 2 domain containing F |
Gene Aliases: |
- |
Location: |
Chr.15:45167214-45201175 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SHF | NM_001301168.1 | 6 | 1677 | NP_001288097.1 |
| NM_001301169.1 | 6 | 1675 | NP_001288098.1 | |
| NM_001301170.1 | 4 | 1385 | NP_001288099.1 | |
| NM_001301171.1 | 6 | 1264 | NP_001288100.1 | |
| NM_138356.2 | 8 | 2102 | NP_612365.2 | |
| XM_006720742.3 | 7 | 1815 | XP_006720805.2 | |
| XM_011522176.2 | 5 | 1580 | XP_011520478.1 | |
| XM_011522177.2 | 5 | 1504 | XP_011520479.1 | |
| XM_017022710.1 | 5 | 1526 | XP_016878199.1 | |
| XM_017022711.1 | 7 | 1928 | XP_016878200.1 | |
| AK096267.1 | 8 | 2101 | ||
| BC007586.1 | 5 | 1039 | AAH07586.1 | |
| BM669861.1 | ||||
| BX416415.2 | 1 | 222 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv518213 | Chr.15:45129560 - 45172570 on Build GRCh38 | Gain |
|
| nsv569312 | Chr.15:45089800 - 45168224 on Build GRCh38 | Loss |
|
| nsv569315 | Chr.15:45153958 - 45480250 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map