Assay Details
Target Gene Details
Entrez Gene ID: | 23464 |
Gene Name: | glycine C-acetyltransferase |
Gene Aliases: |
KBL |
Location: |
Chr.22:37807893-37817177 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GCAT | NM_001171690.1 | NP_001165161.1 | ||
| NM_014291.3 | NP_055106.1 | |||
| XM_005261409.3 | XP_005261466.1 | |||
| XM_005261410.3 | XP_005261467.1 | |||
| XM_005261411.1 | XP_005261468.1 | |||
| XM_011530023.1 | XP_011528325.1 | |||
| XM_017028674.1 | XP_016884163.1 | |||
| XM_017028675.1 | XP_016884164.1 | |||
| XM_017028676.1 | XP_016884165.1 | |||
| XM_017028677.1 | XP_016884166.1 | |||
| AF077740.1 | AAC27720.1 | |||
| AK123190.1 | BAC85552.1 | |||
| AK290093.1 | ||||
| BC014457.1 | AAH14457.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758838 | Chr.22:37607870 - 37869871 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map