Assay Details
Target Gene Details
Entrez Gene ID: | 389123 |
Gene Name: | IQ motif containing F2 |
Gene Aliases: |
- |
Location: |
Chr.3:51861629-51863424 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IQCF2 | NM_203424.1 | NP_982248.1 | ||
| AK128883.1 | ||||
| BC040047.1 | AAH40047.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3817 | Chr.3:51843923 - 51877041 on Build GRCh38 | Insertion |
|
| nsv1001350 | Chr.3:51826801 - 51865950 on Build GRCh38 | Gain |
|
| nsv520211 | Chr.3:51738331 - 52132131 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map