Assay Details
Target Gene Details
Entrez Gene ID: | 26090 |
Gene Name: | abhydrolase domain containing 12 |
Gene Aliases: |
ABHD12A, BEM46L2, C20orf22, PHARC, dJ965G21.2 |
Location: |
Chr.20:25294743-25390982 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABHD12 | NM_015600.4 | NP_056415.1 | ||
| BC014049.2 | AAH14049.1 |
Target Gene Details
Entrez Gene ID: | 5834 |
Gene Name: | phosphorylase, glycogen; brain |
Gene Aliases: |
GPBB |
Location: |
Chr.20:25248070-25298012 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PYGB | NM_002862.3 | 20 | 2761 | NP_002853.2 |
| AB209080.1 | 20 | 2719 | BAD92317.1 | |
| AF432222.1 | 5 | 729 | AAL99928.1 | |
| AK299697.1 | 20 | 2685 | ||
| BC017045.2 | 20 | 2736 | AAH17045.1 | |
| BC030795.2 | 20 | 2737 | AAH30795.1 | |
| J03544.1 | AAA59597.1 | |||
| U47025.1 | 20 | 2686 | AAB60395.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv779e214 | Chr.20:25104610 - 25367702 on Build GRCh38 | Gain |
|
| nsv522873 | Chr.20:23923336 - 26134874 on Build GRCh38 | Gain |
|
| nsv585746 | Chr.20:25090818 - 25489420 on Build GRCh38 | Gain |
|
| dgv4290n100 | Chr.20:25086650 - 25494993 on Build GRCh38 | Gain |
|
| esv3645582 | Chr.20:25284710 - 25318462 on Build GRCh38 | Loss |
|
| nsv833948 | Chr.20:25251090 - 25429904 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map