Assay Details
Target Gene Details
Entrez Gene ID: | 201164 |
Gene Name: | phospholipase D family member 6 |
Gene Aliases: |
ZUC |
Location: |
Chr.17:17200990-17206332 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLD6 | NM_178836.3 | 2 | 2311 | NP_849158.2 |
| XM_017024310.1 | 2 | 2307 | XP_016879799.1 | |
| AK090899.1 | 2 | 2288 | BAC03541.1 | |
| AK098419.1 | 1 | 1430 | BAC05303.1 | |
| BC031263.1 | 2 | 2297 | AAH31263.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833380 | Chr.17:17185102 - 17337892 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map