Assay Details
Target Gene Details
Entrez Gene ID: | 379 |
Gene Name: | ADP ribosylation factor like GTPase 4D |
Gene Aliases: |
ARF4L, ARL6 |
Location: |
Chr.17:43398985-43401136 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARL4D | NM_001661.3 | 2 | 728 | NP_001652.2 |
| XM_011524782.2 | 2 | 656 | XP_011523084.1 | |
| AB060692.1 | 2 | 704 | BAB91080.1 | |
| AK314951.1 | 2 | 720 | ||
| BC000043.2 | 2 | 708 | AAH00043.1 | |
| L38490.1 | 2 | 703 | AAA57126.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2761959 | Chr.17:43322294 - 43565935 on Build GRCh38 | Gain |
|
| nsv833452 | Chr.17:43333582 - 43409552 on Build GRCh38 | Loss |
|
| nsv470588 | Chr.17:43346885 - 43616240 on Build GRCh38 | Gain |
|
| esv3640625 | Chr.17:43361371 - 43577834 on Build GRCh38 | Gain |
|
| nsv833451 | Chr.17:43329618 - 43499146 on Build GRCh38 | Loss |
|
| nsv1065802 | Chr.17:43338154 - 43584333 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map