Assay Details
Target Gene Details
Entrez Gene ID: | 114792 |
Gene Name: | kelch like family member 32 |
Gene Aliases: |
BKLHD5, KIAA1900, UG0030H05, dJ21F7.1 |
Location: |
Chr.6:96898083-97142949 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLHL32 | XM_005266813.4 | 9 | 4741 | XP_005266870.1 |
| XM_017010228.1 | 8 | 4525 | XP_016865717.1 | |
| XM_017010229.1 | 9 | 4698 | XP_016865718.1 |
Target Gene Details
Entrez Gene ID: | 253714 |
Gene Name: | MMS22 like, DNA repair protein |
Gene Aliases: |
C6orf167, dJ39B17.2 |
Location: |
Chr.6:97142161-97283437 on Build GRCh38 |
Assay Gene Location: | Within Exon 31 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MMS22L | NM_198468.2 | 25 | 8521 | NP_940870.2 |
| XM_006715430.1 | 25 | 8452 | XP_006715493.1 | |
| CR749603.1 | 5 | 5131 | CAH18398.1 |
More Information
Set Membership: |
|

Genomic Map