Assay Details
Target Gene Details
Entrez Gene ID: | 254268 |
Gene Name: | AKNA domain containing 1 |
Gene Aliases: |
C1orf62 |
Location: |
Chr.1:108815898-108858242 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AKNAD1 | NM_152763.4 | NP_689976.2 | ||
| NR_049760.1 | ||||
| AK095517.1 | BAC04565.1 | |||
| AL832216.1 | ||||
| BC150642.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2287 | Chr.1:108798077 - 108840621 on Build GRCh38 | Deletion |
|
| nsv547416 | Chr.1:108813740 - 108829252 on Build GRCh38 | Loss |
|
| nsv1013192 | Chr.1:108803093 - 108868275 on Build GRCh38 | Gain |
|

Genomic Map