Assay Details
Target Gene Details
Entrez Gene ID: | 7529 |
Gene Name: | tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein beta |
Gene Aliases: |
GW128, HEL-S-1, HS1, KCIP-1, YWHAA |
Location: |
Chr.20:44885599-44908532 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| YWHAB | NM_003404.4 | 7 | 2292 | NP_003395.1 |
| NM_139323.3 | 6 | 2197 | NP_647539.1 | |
| XM_017028039.1 | 7 | 2096 | XP_016883528.1 | |
| AF107406.1 | 1 | 1057 | AAD44524.1 | |
| AL834170.1 | 6 | 2076 | ||
| EU794580.1 | 7 | 2188 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv954044 | Chr.20:44885960 - 44910159 on Build GRCh38 | Duplication |
|
| nsv586077 | Chr.20:44897496 - 44942370 on Build GRCh38 | Gain |
|
| nsv1058850 | Chr.20:44906262 - 44934643 on Build GRCh38 | Gain |
|
| nsv1062628 | Chr.20:44870663 - 44920098 on Build GRCh38 | Gain |
|
| nsv1056102 | Chr.20:44673341 - 45636248 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map