Assay Details
Target Gene Details
Entrez Gene ID: | 8473 |
Gene Name: | O-linked N-acetylglucosamine (GlcNAc) transferase |
Gene Aliases: |
HINCUT-1, HRNT1, O-GLCNAC |
Location: |
Chr.X:71533062-71575897 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 15 - Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OGT | NM_181672.2 | NP_858058.1 | ||
| NM_181673.2 | NP_858059.1 | |||
| XM_017029907.1 | XP_016885396.1 | |||
| XM_017029908.1 | XP_016885397.1 | |||
| AF223393.1 | AAF31458.1 | |||
| AK313530.1 | ||||
| AL050366.1 | CAB62528.1 | |||
| AL833085.2 | CAD89970.1 | |||
| BC014434.1 | AAH14434.1 | |||
| BC038180.1 | AAH38180.1 | |||
| BX537844.1 | CAD97853.1 | |||
| U77413.1 | AAB63466.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33082 | Chr.X:70420685 - 71888183 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map