Assay Details
Target Gene Details
Entrez Gene ID: | 1353 |
Gene Name: | COX11, cytochrome c oxidase copper chaperone |
Gene Aliases: |
COX11P |
Location: |
Chr.17:54951898-54971024 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COX11 | NR_027941.2 | 5 | 1116 | |
| NR_027942.2 | 5 | 1102 | ||
| NR_135677.1 | 5 | 2705 | ||
| XM_017024194.1 | 6 | 6903 | XP_016879683.1 | |
| XM_017024195.1 | 6 | 6907 | XP_016879684.1 | |
| XM_017024196.1 | 6 | 1196 | XP_016879685.1 | |
| AF044321.1 | 5 | 2572 | AAD08645.1 | |
| AI500638.1 | 1 | 105 | ||
| AI828544.1 | 1 | 87 | ||
| AK124809.1 | 1 | 2892 | ||
| BM975167.1 | 1 | 105 |
Target Gene Details
Entrez Gene ID: | 10040 |
Gene Name: | target of myb1 like 1 membrane trafficking protein |
Gene Aliases: |
SRCASM |
Location: |
Chr.17:54900691-54961967 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TOM1L1 | NM_001321174.1 | NP_001308103.1 | ||
| NM_001321175.1 | NP_001308104.1 | |||
| NM_001321176.1 | NP_001308105.1 | |||
| NM_005486.2 | NP_005477.2 | |||
| XM_017024002.1 | XP_016879491.1 | |||
| AJ010071.1 | CAA08993.1 | |||
| AK001893.1 | ||||
| AK223125.1 | BAD96845.1 | |||
| AK300823.1 | ||||
| AK303912.1 | ||||
| AK303913.1 | ||||
| AK315039.1 | ||||
| AK315907.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv3266n100 | Chr.17:54817463 - 55002289 on Build GRCh38 | Loss |
|
| nsv457853 | Chr.17:54910871 - 54982748 on Build GRCh38 | Loss |
|
| esv3640830 | Chr.17:54919823 - 54998005 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map