Assay Details
Target Gene Details
Entrez Gene ID: | 128954 |
Gene Name: | GRB2 associated binding protein family member 4 |
Gene Aliases: |
- |
Location: |
Chr.22:16961936-17008281 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GAB4 | NM_001037814.1 | NP_001032903.1 | ||
| XM_011546114.2 | XP_011544416.1 | |||
| XM_011546115.1 | XP_011544417.1 | |||
| XM_011546116.2 | XP_011544418.1 | |||
| XM_017028575.1 | XP_016884064.1 | |||
| AB076978.1 | BAD06454.1 | |||
| AK057252.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv4460n100 | Chr.22:16384224 - 17135620 on Build GRCh38 | Gain |
|
| esv3647204 | Chr.22:16939332 - 17123705 on Build GRCh38 | Gain |
|
| dgv4463n100 | Chr.22:16749141 - 17081023 on Build GRCh38 | Gain |
|
| nsv1067241 | Chr.22:16757380 - 17351252 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map