Assay Details
Target Gene Details
Entrez Gene ID: | 3183 |
Gene Name: | heterogeneous nuclear ribonucleoprotein C (C1/C2) |
Gene Aliases: |
C1, C2, HNRNP, HNRPC, SNRPC |
Location: |
Chr.14:21209136-21269479 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv974313 | Chr.14:21210389 - 21211709 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map