Assay Details
Target Gene Details
Entrez Gene ID: | 5360 |
Gene Name: | phospholipid transfer protein |
Gene Aliases: |
BPIFE, HDLCQ9 |
Location: |
Chr.20:45898620-45912364 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLTP | NM_001242920.1 | 7 | 805 | NP_001229849.1 |
| NM_001242921.1 | 7 | 717 | NP_001229850.1 | |
| NM_006227.3 | 9 | 1090 | NP_006218.1 | |
| NM_182676.2 | 8 | 934 | NP_872617.1 | |
| AB076694.1 | 9 | 874 | BAB79630.1 | |
| AF052157.1 | 6 | 592 | ||
| AK092397.1 | 9 | 831 | ||
| AK092523.1 | 8 | 722 | ||
| AK097009.1 | 8 | 1388 | ||
| AK222690.1 | 9 | 878 | BAD96410.1 | |
| AK289371.1 | 8 | 722 | ||
| AK293150.1 | 7 | 633 | ||
| AK299181.1 | 7 | 597 | ||
| BC005045.1 | 8 | 715 | AAH05045.1 | |
| BC019847.1 | 9 | 872 | AAH19847.1 | |
| BC019898.1 | 9 | 865 | AAH19898.1 | |
| L26232.1 | 9 | 880 | AAA36443.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3393 | Chr.20:45903113 - 45913226 on Build GRCh38 | Deletion |
|
| nsv517296 | Chr.20:45854586 - 45965936 on Build GRCh38 | Loss |
|
| nsv833991 | Chr.20:45885631 - 46068708 on Build GRCh38 | Loss |
|
| nsv586134 | Chr.20:45899835 - 45905120 on Build GRCh38 | Gain |
|
| nsv833990 | Chr.20:45771510 - 45946844 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map