Assay Details
Target Gene Details
Entrez Gene ID: | 81608 |
Gene Name: | factor interacting with PAPOLA and CPSF1 |
Gene Aliases: |
FIP1, Rhe, hFip1 |
Location: |
Chr.4:53377572-53462611 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
Target Gene Details
Entrez Gene ID: | 84708 |
Gene Name: | ligand of numb-protein X 1 |
Gene Aliases: |
LNX, MPDZ, PDZRN2 |
Location: |
Chr.4:53458264-53652524 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LNX1 | XM_005265785.4 | 11 | 4148 | XP_005265842.1 |
| XM_017008776.1 | 12 | 4209 | XP_016864265.1 | |
| AF237782.1 | 10 | 3652 | AAK15039.1 |
More Information
Set Membership: |
|

Genomic Map