Assay Details
Target Gene Details
Entrez Gene ID: | 54902 |
Gene Name: | tetratricopeptide repeat domain 19 |
Gene Aliases: |
2010204O13Rik, MC3DN2 |
Location: |
Chr.17:15999380-16045428 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC19 | NM_001271420.1 | NP_001258349.1 | ||
| NM_017775.3 | NP_060245.3 | |||
| XM_017024801.1 | XP_016880290.1 | |||
| XM_017024802.1 | XP_016880291.1 | |||
| AK000350.1 | BAA91103.1 | |||
| AK055780.1 | ||||
| AK056878.1 | ||||
| AK297783.1 | ||||
| BC011698.2 | AAH11698.2 | |||
| BC073796.1 | AAH73796.1 | |||
| BC105128.1 | AAI05129.1 | |||
| BC112107.1 | AAI12108.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv3108n100 | Chr.17:15966243 - 16193927 on Build GRCh38 | Gain |
|
| esv3892973 | Chr.17:15947882 - 16030711 on Build GRCh38 | Gain |
|
| esv3572431 | Chr.17:15951820 - 16194089 on Build GRCh38 | Gain |
|
| dgv3107n100 | Chr.17:15703953 - 16150198 on Build GRCh38 | Gain |
|
| esv2715702 | Chr.17:15750514 - 16668268 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map