Assay Details
Target Gene Details
Entrez Gene ID: | 92162 |
Gene Name: | transmembrane protein 88 |
Gene Aliases: |
- |
Location: |
Chr.17:7854886-7856099 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM88 | NM_001319941.1 | NP_001306870.1 | ||
| NM_203411.1 | NP_981956.1 | |||
| XM_005256856.3 | XP_005256913.1 | |||
| AK000032.1 | ||||
| BC057812.1 | AAH57812.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv510n67 | Chr.17:7852429 - 7911448 on Build GRCh38 | Gain |
|
| nsv833351 | Chr.17:7755691 - 7893863 on Build GRCh38 | Loss |
|
| nsv1070804 | Chr.17:7853581 - 7856982 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map