Assay Details
Target Gene Details
Entrez Gene ID: | 2321 |
Gene Name: | fms related tyrosine kinase 1 |
Gene Aliases: |
FLT, FLT-1, VEGFR-1, VEGFR1 |
Location: |
Chr.13:28300346-28495128 on Build GRCh38 |
Assay Gene Location: | Within Exon 33 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FLT1 | NM_002019.4 | 30 | 4248 | NP_002010.2 |
| XM_017020485.1 | 29 | 4134 | XP_016875974.1 | |
| AB209050.1 | 14 | 1604 | BAD92287.1 | |
| AF063657.2 | 30 | 3963 | AAC16449.2 | |
| BU151621.1 | 7 | 786 | ||
| DQ836394.1 | 16 | 1788 | ||
| DQ836395.1 | 17 | 1850 | ||
| EF491868.1 | 12 | 1497 | ||
| EF491869.1 | 9 | 1284 | ||
| EF491870.1 | 10 | 1188 | ||
| JF509744.1 | 12 | 1381 | ||
| JF509745.1 | 3 | 339 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34052 | Chr.13:28074532 - 28352440 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map