Assay Details
Target Gene Details
Entrez Gene ID: | 389136 |
Gene Name: | vestigial like family member 3 |
Gene Aliases: |
VGL-3, VGL3 |
Location: |
Chr.3:86937969-86991293 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VGLL3 | NM_001320494.1 | 4 | 9763 | NP_001307423.1 |
| NM_016206.3 | 4 | 9922 | NP_057290.2 | |
| XM_006713138.3 | 4 | 10089 | XP_006713201.1 | |
| AF099505.1 | 1 | 1058 | AAF00091.1 | |
| AK095826.1 | 1 | 2306 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv527279 | Chr.3:86796784 - 87814130 on Build GRCh38 | Gain |
|
| esv3596879 | Chr.3:86908792 - 86940260 on Build GRCh38 | Loss |
|
| nsv519148 | Chr.3:86781927 - 86945974 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map