Assay Details
Target Gene Details
Entrez Gene ID: | 1912 |
Gene Name: | polyhomeotic homolog 2 |
Gene Aliases: |
EDR2, HPH2, PH2 |
Location: |
Chr.1:33323623-33431017 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 20 - Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PHC2 | NM_004427.3 | NP_004418.2 | ||
| NM_198040.2 | NP_932157.1 | |||
| XM_005270570.1 | XP_005270627.1 | |||
| XM_017000513.1 | XP_016856002.1 | |||
| XM_017000514.1 | XP_016856003.1 | |||
| XM_017000515.1 | XP_016856004.1 | |||
| XM_017000516.1 | XP_016856005.1 | |||
| XM_017000517.1 | XP_016856006.1 | |||
| XM_017000518.1 | XP_016856007.1 | |||
| XM_017000519.1 | XP_016856008.1 | |||
| XM_017000520.1 | XP_016856009.1 | |||
| XM_017000521.1 | XP_016856010.1 | |||
| AJ419231.1 | CAD11673.1 | |||
| AK056429.1 | ||||
| AK128821.1 | BAC87622.1 | |||
| AK292905.1 | ||||
| AK294426.1 | ||||
| BC028396.1 | ||||
| BC029269.1 | AAH29269.1 | |||
| BC068573.1 | AAH68573.1 | |||
| BC092492.1 | AAH92492.1 | |||
| BC110863.1 | ||||
| BC130630.1 | ||||
| BC144121.1 | ||||
| U89278.1 | AAC51170.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522364 | Chr.1:33266591 - 33367332 on Build GRCh38 | Loss |
|
| nsv516693 | Chr.1:33319026 - 33354533 on Build GRCh38 | Loss |
|
| nsv545963 | Chr.1:33254041 - 33354533 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map